accessibility menu, dialog, popup

UserName

OOOOH

CAS RN 79831-88-2

IUPAC Name: [3,4,5-tris(benzyloxy)phenyl]methanol
Synonyms: [3,4,5-tris(benzyloxy)phenyl]methanol
Molecular Weight (g/mol): 426.51
Molecular Formula: C28H26O4
InChi Key: FWPHSYJOSSBILN-UHFFFAOYSA-N
SMILES: OCC1=CC(OCC2=CC=CC=C2)=C(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results