accessibility menu, dialog, popup

UserName

NNOO

CAS RN 79463-77-7

IUPAC Name: cyano(diphenoxymethylidene)amine
Synonyms: cyano(diphenoxymethylidene)amine
Molecular Weight (g/mol): 238.25
Molecular Formula: C14H10N2O2
InChi Key: SLIKWVTWIGHFJE-UHFFFAOYSA-N
SMILES: N#CN=C(OC1=CC=CC=C1)OC1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)