accessibility menu, dialog, popup

UserName

CAS RN 79277-27-3

CH3OOSSOOHNOHNNCH3NOH3CN

CAS RN 79277-27-3

IUPAC Name: methyl 3-({[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)carbamoyl]amino}sulfonyl)thiophene-2-carboxylate
Synonyms: harmony
Molecular Weight (g/mol): 387.39
Molecular Formula: C12H13N5O6S2
InChi Key: AHTPATJNIAFOLR-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C=CS1)S(=O)(=O)NC(=O)NC1=NC(C)=NC(OC)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results