accessibility menu, dialog, popup

UserName

CH3H3CBrOHBrBrOHBr

CAS RN 79-94-7

IUPAC Name: 2,6-dibromo-4-[2-(3,5-dibromo-4-hydroxyphenyl)propan-2-yl]phenol
Synonyms: tetrabromobisphenol A saytex RB 100PC tetrabromodiphenylopropane firemaster BP 4A fire guard 2000 tbbpa cpd great lakes BA-59P tetrabromodian bromdian
Molecular Weight (g/mol): 543.88
Molecular Formula: C15H12Br4O2
InChi Key: VEORPZCZECFIRK-UHFFFAOYSA-N
SMILES: CC(C)(C1=CC(Br)=C(O)C(Br)=C1)C1=CC(Br)=C(O)C(Br)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)