accessibility menu, dialog, popup

UserName

H3COCH3O

CAS RN 79-20-9

IUPAC Name: methyl acetate
Synonyms: methyl acetate
Molecular Weight (g/mol): 74.08
Molecular Formula: C3H6O2
InChi Key: KXKVLQRXCPHEJC-UHFFFAOYSA-N
SMILES: COC(C)=O
Percent Purity 
  • (1)
  • (4)
  • (4)
  • (4)
  • (3)
Quantity 
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (3)
  • (2)
  • (4)
  • (4)
Molecular Weight (g/mol) 
  • (4)
  • (5)
Boiling Point 
  • (2)
  • (1)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (8)
  • (8)
  • (1)
  • (1)
  • (7)
  • (1)
  • (4)
  • (4)
  • (4)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)
  • (4)
  • (4)
  • (5)
  • (2)
  • (1)
  • (5)
  • (3)
  • (3)
  • (1)