accessibility menu, dialog, popup

UserName

NH2OCl

CAS RN 79-07-2

IUPAC Name: 2-chloroacetamide
Synonyms: chloroacetamide
Molecular Weight (g/mol): 93.51
Molecular Formula: C2H4ClNO
InChi Key: VXIVSQZSERGHQP-UHFFFAOYSA-N
SMILES: NC(=O)CCl
Boiling Point 
  • (2)
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (4)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (5)
  • (2)
  • (3)
  • (4)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (7)