missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroacetamide 98.0+%, TCI America™

Catalog No. C0086500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0086500G 500 g
C008625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0086500G Supplier TCI America Supplier No. C0086500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79-07-2
Molecular Formula C2H4ClNO
Molecular Weight (g/mol) 93.51
MDL Number MFCD00008027
InChI Key VXIVSQZSERGHQP-UHFFFAOYSA-N
Synonym chloroacetamide, acetamide, 2-chloro, chloracetamide, 2-chloroethanamide, microcide, mergal af, chloracetamid, usaf do-29, 2-chloracetamide, 2-chloro-acetamide
PubChem CID 6580
IUPAC Name 2-chloroacetamide
SMILES NC(=O)CCl

Specifications

Melting Point 120°C
Color White
Boiling Point 225°C
Quantity 500 g
UN Number 2811
Formula Weight 93.51
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloroacetamide
EINECSNumber (2)-1148&(2)-2642
RTECSNumber AB5075000
TSCA Yes