accessibility menu, dialog, popup

UserName

CAS RN 78338-22-4

IUPAC Name: 3,7-diamino-5λ⁴-phenothiazin-5-ylium
Synonyms: thionine(1+) thionine cation
Molecular Weight (g/mol): 228.29
Molecular Formula: C12H10N3S
InChi Key: CBBMGBGDIPJEMI-UHFFFAOYSA-N
SMILES: NC1=CC2=[S+]C3=CC(N)=CC=C3N=C2C=C1
Quantity 
  • (2)
  • (2)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)