missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thionin acetate, pure, high purity biological stain

Catalog No. AC229840250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC229840250 25 g Glass bottle
AC229840050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC229840250 Supplier Thermo Scientific Chemicals Supplier No. 229840250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 78338-22-4 | C14H13N3O2S | 287.337 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 78338-22-4
Molecular Formula C14H13N3O2S
Molecular Weight (g/mol) 287.337
MDL Number MFCD00081194
InChI Key JOIRQYHDJINFGA-UHFFFAOYSA-N
Synonym 3, 7-Diaminophenothiazin-5-ium acetate
PubChem CID 2724414
IUPAC Name (7-aminophenothiazin-3-ylidene)azanium;acetate
SMILES CC(=O)[O-].C1=CC2=C(C=C1N)SC3=CC(=[NH2+])C=CC3=N2

Specifications

Dye Content 85% min.
Infrared Spectrum Authentic
Lambda Max 597 to 601nm (in water)
Packaging Glass bottle
UV Spectrometry Complies
Beilstein 27, 392
Merck Index 14, 9346
Color Green
Solubility Information Solubility in water: ca 2.5g/L (25°C). Other solubilities: practically insoluble in ethanol
Quantity 25 g
Physical Form Crystalline Powder or Crystals
Formula Weight 287.33
Grade Pure
Chemical Name or Material Thionin acetate, Pure
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only