missing translation for 'onlineSavingsMsg'
Learn More

Thionin acetate

Catalog No. AAA1891214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1891214 25 g
AAA1891206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1891214 Supplier Thermo Scientific Chemicals Supplier No. A1891214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used as a biogical stain | CAS: 78338-22-4 | C14H13N3O2S | 287.337 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Thionin acetate is a strongly staining metachromatic dye that is widely used for biological staining. Thionine can also be used in place of Schiff reagent in quantitative Feulgen staining of DNA. It can also be used to mediate electron transfer in microbial fuel cells. It is also employed as a general nuclear stain for chromatin and mucin; for staining Nissl granules by end-point buffering and for showing malignant cells in frozen biopsy sections; and as a stain for semi-thin sections of plant material for light microscopy.

Solubility
Soluble in water (∼2.5 mg/ml at 25°C), and HOAc: H2O (1:1) (1 mg/ml). Insoluble in ethanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 78338-22-4
Molecular Formula C14H13N3O2S
Molecular Weight (g/mol) 287.337
MDL Number MFCD00081194
InChI Key JOIRQYHDJINFGA-UHFFFAOYSA-N
PubChem CID 2724414
IUPAC Name (7-aminophenothiazin-3-ylidene)azanium;acetate
SMILES CC(=O)[O-].C1=CC2=C(C=C1N)SC3=CC(=[NH2+])C=CC3=N2

Specifications

Solubility Information Soluble in water (∽2.5mg/ml at 25°C),and HOAc: H2O (1:1) (1mg/ml). Insoluble in ethanol.
Quantity 25 g
Formula Weight 287.34
Chemical Name or Material Thionin acetate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only