accessibility menu, dialog, popup

UserName

CH3I

CAS RN 782477-81-0

IUPAC Name: 4-iodo-4'-propyl-1,1'-biphenyl
Synonyms: 4-iodo-4'-propyl-1,1'-biphenyl
Molecular Weight (g/mol): 322.19
Molecular Formula: C15H15I
InChi Key: MKDLYSABBHXXSV-UHFFFAOYSA-N
SMILES: CCCC1=CC=C(C=C1)C1=CC=C(I)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results