accessibility menu, dialog, popup

UserName

CH3H3CO

CAS RN 78-84-2

IUPAC Name: 2-methylpropanal
Synonyms: isobutyraldehyde valine aldehyde isobutyraldehyd isobutylaldehyde isobutyryl aldehyde isobutyraldehyde (natural) 2-methylpropanal (substance)
Molecular Weight (g/mol): 72.11
Molecular Formula: C4H8O
InChi Key: AMIMRNSIRUDHCM-UHFFFAOYSA-N
SMILES: CC(C)C=O
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)