missing translation for 'onlineSavingsMsg'
Learn More

Isobutyraldehyde, ≥97% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019487
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019487 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019487 Supplier Merck Emd Millipore Supplier No. 685291ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 78-84-2
Ignition Point 384°F
Vapor Pressure 66 mmHg (4.4°C)
Vapor Density 2.5 (vs. air)
Density 0.79 g/mL (at 25°C)
Physical Form Neat
Linear Formula (CH3)2 CHCHO
MDL Number MFCD00006980
Melting Point -65°C
Boiling Point 63°C
Synonym 2-Methylpropanal; 2-Methylpropionaldehyde
Type Analytical Standard
Quantity 1 mL
Formula Weight 72.11
Percent Purity ≥97% (GC)
Grade Analytical Standard
Show More Show Less