accessibility menu, dialog, popup

UserName

CAS RN 78-40-0

IUPAC Name: triethyl phosphate
Synonyms: triethyl phosphate|ethyl phosphate|triethylphosphate|TEP
Molecular Weight (g/mol): 182.16
Molecular Formula: C6H15O4P
InChi Key: DQWPFSLDHJDLRL-UHFFFAOYSA-N
SMILES: CCOP(=O)(OCC)OCC
Boiling Point 
  • (7)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (9)
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (2)
  • (7)
  • (7)
  • (1)
  • (2)
  • (1)
  • (9)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (7)