missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium metaperiodate, 99.0%, for analysis EMSURE™ ACS, Reag. Ph Eur, MilliporeSigma™
Specifications
Specifications
| Melting Point | 270°C (decomposes) |
| pH | 5.2 (50 g/L, H2O, 20°C) |
| Physical Form | Crystals |
| Quantity | 250 g |
| Packaging | Plastic Bottle |
| Assay Percent Range | Assay (iodometric): ≥ 99.0%; Assay (out of dried substance): 99.8 to 100.3% |
| Density | 3.87 g/cm3 (at 20°C); Bulk Density: 2900 kg/m3 |
| Solubility Information | 91 g/L |
| Percent Purity | ≥99.0% |
| Grade | ACS |
| Show More |
Safety and Handling
| Recommended Storage | Store at 5 to 30°C |