missing translation for 'onlineSavingsMsg'
Learn More

Sodium (Meta)Periodate, Solstice

Catalog No. 60045816
Change view
Click to view available options
Quantity:
100 g
500 g
6 x 500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60045816 6 x 500 g
6001370 100 g
6001371 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60045816 Supplier Solstice Supplier No. 303236X500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99.8%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7790-28-5
Molecular Formula INaO4
Molecular Weight (g/mol) 213.89
MDL Number MFCD00003534
InChI Key JQWHASGSAFIOCM-UHFFFAOYSA-M
Synonym sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate
PubChem CID 23667635
ChEBI CHEBI:75226
IUPAC Name sodium;periodate
SMILES [Na+].[O-][I](=O)(=O)=O

Specifications

Physical Form Crystalline
Quantity 6 x 500 g
Linear Formula NaIO4
UN Number UN3085
Formula Weight 213.89
Percent Purity ≥99.8%
Grade ACS/puriss. p.a.
Chemical Name or Material Sodium (Meta)Periodate, Reag. Ph. Eur. reag. ISO
Hazard Statement P220-P273-P280-P305 + P351 + P338-P310
Precautionary Statement H271-H314-H372-H400
DOTInformation class 5.1 - PG 1 - acidic - EHS - Oxidizing solid, corrosive, n.o.s., HI: all
EINECSNumber 232-197-6
RTECSNumber SD4550000