accessibility menu, dialog, popup

UserName

MgClCl

CAS RN 7786-30-3

IUPAC Name: magnesium(2+) dichloride
Synonyms: magnesium chloride|MgCl2|magnesium chloride anhydrous|magnesium dichloride|magnesiumchlorid
Molecular Weight (g/mol): 95.21
Molecular Formula: Cl2Mg
InChi Key: TWRXJAOTZQYOKJ-UHFFFAOYSA-L
SMILES: [Mg++].[Cl-].[Cl-]
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (3)
  • (5)
  • (5)
Grade 
  • (2)
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (2)
  • (1)
  • (15)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (5)
  • (5)
  • (2)
  • (4)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)