missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chloride, ultra dry, 99.99% (metals basis)

Catalog No. AA4285022
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4285022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4285022 Supplier Thermo Scientific Chemicals Supplier No. 04285022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7786-30-3 | Cl2Mg | 95.21 g/mol

Magnesium chloride is generally used as a Lewis acid catalyst. It is used as a flocculating agent, analytical reagent, anesthetic for cephalopods, and also in various magnesium cements, and ceramics. It is used for dust control, and road stabilization. It is used in fire extinguishers, polymerase chain reaction (PCR) and hydrogen storage. It is a precursor for magnesium metal and other magnesium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7786-30-3
Molecular Formula Cl2Mg
Molecular Weight (g/mol) 95.21
MDL Number MFCD00011106
InChI Key TWRXJAOTZQYOKJ-UHFFFAOYSA-L
Synonym magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride
PubChem CID 5360315
ChEBI CHEBI:6636
SMILES [Mg++].[Cl-].[Cl-]

Specifications

Melting Point 714°C
Color White to Yellow
Physical Form Powder
Quantity 100 g
Odor Odorless
Sensitivity Hygroscopic
Appearance White to yellow powder
Assay Percent Range 99.99% (Metal basis)
Density 2.32
Solubility Information Highly soluble in water with evolution of heat. Slightly soluble in acetone and pyridine.
IUPAC Name magnesium;dichloride
Formula Weight 95.22
Chemical Name or Material Magnesium chloride, ultra dry
Show More Show Less
Hazard Category H303: May be harmful if swallowed.
EINECSNumber 232-094-6
RTECSNumber OM2800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only