accessibility menu, dialog, popup

UserName

SbFFF

CAS RN 7783-56-4

IUPAC Name: trifluorostibane
Synonyms: antimony trifluoride
Molecular Weight (g/mol): 178.76
Molecular Formula: F3Sb
InChi Key: GUNJVIDCYZYFGV-UHFFFAOYSA-K
SMILES: F[Sb](F)F
Molecular Weight (g/mol) 
  • (7)
  • (1)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (7)
  • (7)
  • (1)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)