missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) fluoride, 98%

Catalog No. AAA1406830
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1406830 250 g
AAA1406818 50 g
AAA140680B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1406830 Supplier Thermo Scientific Chemicals Supplier No. A1406830

CAS: 7783-56-4 | F3Sb | 178.755 g/mol

Antimony(III) fluoride is used as a reagent in inorganic and organofluorine chemistry. It acts as a fluorination reagent and electroplating agent. Further, it is used in the preparation of dye, pottery, porcelains, ceramic enamels and glazes. It is utilized in the preparation of freon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Antimony(III) fluoride is used as a reagent in inorganic and organofluorine chemistry. It acts as a fluorination reagent and electroplating agent. Further, it is used in the preparation of dye, pottery, porcelains, ceramic enamels and glazes. It is utilized in the preparation of freon.

Solubility
Soluble in water, methanol and acetone. Insoluble in ammonia.

Notes
Moisture sensitive. It is incompatible with acids, perchloric acid, water and oxidizing agents.

Chemical Identifiers

CAS 7783-56-4
Molecular Formula F3Sb
Molecular Weight (g/mol) 178.76
MDL Number MFCD00011218
InChI Key GUNJVIDCYZYFGV-UHFFFAOYSA-K
Synonym antimony trifluoride, trifluorostibine, antimony fluoride, antimony iii fluoride, stibine, trifluoro, trifluoroantimony, antimoine fluorure, antimonous fluoride, antimoine fluorure french, hsdb 438
PubChem CID 24554
IUPAC Name trifluorostibane
SMILES F[Sb](F)F

Specifications

Boiling Point 376°C
Melting Point 292°C
Quantity 250 g
UN Number UN2923
Merck Index 14,709
Solubility Information Soluble in water,methanol and acetone. Insoluble in ammonia.
Formula Weight 178.75
Percent Purity 98%
Odor Odorless
Sensitivity Moisture sensitive
Density 4.379
Chemical Name or Material Antimony(III) fluoride
Show More Show Less
Hazard Category H301+H311+H331-H314-H335
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 232-009-2
RTECSNumber CC5150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only