accessibility menu, dialog, popup

UserName

CH3SiH3C

CAS RN 778-24-5

IUPAC Name: dimethyldiphenylsilane
Synonyms: dimethyldiphenylsilane
Molecular Weight (g/mol): 212.37
Molecular Formula: C14H16Si
InChi Key: WJKVFIFBAASZJX-UHFFFAOYSA-N
SMILES: C[Si](C)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results