accessibility menu, dialog, popup

UserName

OH

CAS RN 773-99-9

IUPAC Name: 2-(naphthalen-1-yl)ethan-1-ol
Synonyms: 2-(naphthalen-1-yl)ethanol
Molecular Weight (g/mol): 172.23
Molecular Formula: C12H12O
InChi Key: RXWNCMHRJCOWDK-UHFFFAOYSA-N
SMILES: OCCC1=C2C=CC=CC2=CC=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)