missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Naphthyl)ethanol 95.0+%, TCI America™

Catalog No. N05755G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05755G 5 g
N057525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05755G Supplier TCI America Supplier No. N05755G

Chemical Identifiers

CAS 773-99-9
Molecular Formula C12H12O
Molecular Weight (g/mol) 172.227
MDL Number MFCD00004050
InChI Key RXWNCMHRJCOWDK-UHFFFAOYSA-N
Synonym 1-naphthaleneethanol, 2-1-naphthyl ethanol, 2-naphthalen-1-yl ethanol, 1-2-hydroxyethyl naphthalene, naphthalen-1-ethanol, 2-naphthylethanol, 1-naphthyl ethanol, 1-naphthalene ethanol, 2-naphthalen-1-yl ethan-1-ol, 1-napthaleneethanol
PubChem CID 13047
IUPAC Name 2-naphthalen-1-ylethanol
SMILES C1=CC=C2C(=C1)C=CC=C2CCO

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 179°C
Quantity 5 g
Formula Weight 172.23
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(1-Naphthyl)ethanol