missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Naphthyl)ethanol, 95%, Thermo Scientific Chemicals

Catalog No. AAH2721809
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2721809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2721809 Supplier Thermo Scientific Chemicals Supplier No. H2721809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 773-99-9
Molecular Formula C12H12O
Molecular Weight (g/mol) 172.227
MDL Number MFCD00004050
InChI Key RXWNCMHRJCOWDK-UHFFFAOYSA-N
Synonym 1-naphthaleneethanol, 2-1-naphthyl ethanol, 2-naphthalen-1-yl ethanol, 1-2-hydroxyethyl naphthalene, naphthalen-1-ethanol, 2-naphthylethanol, 1-naphthyl ethanol, 1-naphthalene ethanol, 2-naphthalen-1-yl ethan-1-ol, 1-napthaleneethanol
PubChem CID 13047
IUPAC Name 2-naphthalen-1-ylethanol
SMILES C1=CC=C2C(=C1)C=CC=C2CCO

Specifications

Melting Point 60°C to 64°C
Boiling Point 186°C (17 mmHg)
Quantity 10 g
Formula Weight 172.23
Percent Purity 95%
Chemical Name or Material 2-(1-Naphthyl)ethanol
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 212-260-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only