accessibility menu, dialog, popup

UserName

NH4H3CPOHONH2OO

CAS RN 77182-82-2

IUPAC Name: ammonium 2-amino-4-[hydroxy(methyl)phosphoryl]butanoate
Synonyms: glufosinate-ammonium glufosinate ammonium (iso)
Molecular Weight (g/mol): 198.16
Molecular Formula: C5H15N2O4P
InChi Key: ZBMRKNMTMPPMMK-UHFFFAOYNA-N
SMILES: [NH4+].CP(O)(=O)CCC(N)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results