accessibility menu, dialog, popup

UserName

CH3NOOHOO

CAS RN 77128-73-5

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)-3-phenylpropanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl](methyl)amino}-3-phenylpropanoic acid
Molecular Weight (g/mol): 401.46
Molecular Formula: C25H23NO4
InChi Key: GBROUWPNYVBLFO-UHFFFAOYNA-N
SMILES: CN(C(CC1=CC=CC=C1)C(O)=O)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)