missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-N-methyl-L-phenylalanine, 95%
Description
N-Fmoc-N-methyl-L-phenylalanine is used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Fmoc-N-methyl-L-phenylalanine is used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in DMF. (0.3gram in 2ml)
Notes
Protect from heat. Incompatible with heat and oxidizing agents.
N-Fmoc-N-methyl-L-phenylalanine is used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in DMF. (0.3gram in 2ml)
Notes
Protect from heat. Incompatible with heat and oxidizing agents.
Specifications
Specifications
| Color | White |
| Beilstein | 4768138 |
| Solubility Information | Soluble in DMF. (0.3gram in 2ml) |
| Quantity | 5 g |
| Formula Weight | 401.46 |
| Percent Purity | 95% |
| Physical Form | Powder |
| Chemical Name or Material | N-Fmoc-N-methyl-L-phenylalanine |
Safety and Handling
| TSCA | No |
| storageNote1 | 2° to 8°C |
| Recommended Storage | Keep cold |
RUO – Research Use Only