missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-phenylalanine 98.0+%, TCI America™

Catalog No. F11675G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11675G 5 g
F11671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F11675G Supplier TCI America Supplier No. F11675G
Only null left

Chemical Identifiers

CAS 77128-73-5
Molecular Formula C25H23NO4
Molecular Weight (g/mol) 401.462
MDL Number MFCD00151938
InChI Key GBROUWPNYVBLFO-QHCPKHFHSA-N
Synonym fmoc-n-me-phe-oh, fmoc-n-methyl-l-phenylalanine, fmoc-mephe-oh, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-phenylpropanoic acid, n-fmoc-n-methyl-l-phenylalanine, l-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-phenylalanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-phenylpropanoic acid, fmoc-l-mephe-oh, ambotzfaa1403
PubChem CID 978356
IUPAC Name (2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-phenylpropanoic acid
SMILES CN(C(CC1=CC=CC=C1)C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Melting Point 134°C
Quantity 5 g
Formula Weight 401.46
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-phenylalanine