missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) chloride, MilliporeSigma™

Catalog No. M8039450500
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M8039450500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M8039450500 Supplier Technidata Supplier No. 8.03945.0500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For synthesis

Chemical Identifiers

CAS 7705-08-0
Molecular Formula Cl3Fe
Molecular Weight (g/mol) 162.20
MDL Number MFCD00011005
InChI Key RBTARNINKXHZNM-UHFFFAOYSA-K
Synonym Ferric chloride; Iron trichloride
IUPAC Name iron(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Physical Form Powder
Quantity 500 g
UN Number UN1773
Solubility Information Soluble in water: 920g/L at 25°C
Formula Weight 162.2
Percent Purity ≥98.0%
Grade Not Available
Odor Stinging
Vapor Pressure 1hPa (20°C)
Density 2.89g/cm3 at 25°C
Chemical Name or Material Iron(III) chloride
Show More Show Less
Hazard Category H290: May be corrosive to metals.
Hazard Statement H302: Harmful if swallowed.
Precautionary Statement H315: Causes skin irritation.
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 231-729-4
RTECSNumber LJ9100000
Recommended Storage Store at 15° to 25°C (59° to 77°F), Tightly closed. Dry.