CAS RN 77-06-5
Gibberellic acid, 99%
CAS: 77-06-5 Molecular Formula: C19H22O6 Molecular Weight (g/mol): 346.38 MDL Number: MFCD00079329 InChI Key: IXORZMNAPKEEDV-QTWFBFKQSA-N Synonym: gibberellic acid,gibberellin a3,gibberellin,gibreskol,brellin,cekugib,grocel,gibberellin x,gibberellic acid ga3,gib-tabs PubChem CID: 91757643 IUPAC Name: (1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylic acid SMILES: CC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O
Gibberellic Acid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Gibberellic acid is an endogenous plant growth regulator
Gibberellic acid, 98%
CAS: 77-06-5 Molecular Formula: C19H22O6 Molecular Weight (g/mol): 346.38 MDL Number: MFCD00079329 InChI Key: IXORZMNAPKEEDV-QTWFBFKQSA-N Synonym: gibberellic acid,gibberellin a3,gibberellin,gibreskol,brellin,cekugib,grocel,gibberellin x,gibberellic acid ga3,gib-tabs PubChem CID: 91757643 IUPAC Name: (1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylic acid SMILES: CC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O
Gibberellic acid, 90+%
CAS: 77-06-5 Molecular Formula: C19H22O6 Molecular Weight (g/mol): 346.38 MDL Number: MFCD00079329 InChI Key: IXORZMNAPKEEDV-QTWFBFKQSA-N Synonym: gibberellic acid,gibberellin a3,gibberellin,gibreskol,brellin,cekugib,grocel,gibberellin x,gibberellic acid ga3,gib-tabs PubChem CID: 91757643 IUPAC Name: (1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylic acid SMILES: CC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O
Gibberellic Acid, 90%
CAS: 77-06-5 Molecular Formula: C19H22O6 Molecular Weight (g/mol): 346.38 MDL Number: MFCD00079329 InChI Key: IXORZMNAPKEEDV-QTWFBFKQSA-N Synonym: gibberellic acid,gibberellin a3,gibberellin,gibreskol,brellin,cekugib,grocel,gibberellin x,gibberellic acid ga3,gib-tabs PubChem CID: 91757643 IUPAC Name: (1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylic acid SMILES: CC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O
Gibberellin A3 98.0+%, TCI America™
CAS: 77-06-5 Molecular Formula: C19H22O6 Molecular Weight (g/mol): 346.38 MDL Number: MFCD00079329 InChI Key: IXORZMNAPKEEDV-QTWFBFKQSA-N Synonym: gibberellic acid,gibberellin a3,gibberellin,gibreskol,brellin,cekugib,grocel,gibberellin x,gibberellic acid ga3,gib-tabs PubChem CID: 91757643 IUPAC Name: (1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylic acid SMILES: CC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O