missing translation for 'onlineSavingsMsg'
Learn More

Gibberellic Acid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015487
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015487 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015487 Supplier Merck Emd Millipore Supplier No. PHR1567500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gibberellic acid is an endogenous plant growth regulator

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 77-06-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C19H22O6
MDL Number MFCD00079329
Synonym Gibberellic acid; GA3; Gibberellin A3
Type Certified Reference Material
Quantity 500 mg
Formula Weight 346.37
Grade Certified Reference Material