accessibility menu, dialog, popup

UserName

H3CF

CAS RN 76802-61-4

IUPAC Name: 1-fluoro-4-(4-pentylcyclohexyl)benzene
Synonyms: 1-fluoro-4-(4-pentylcyclohexyl)benzene
Molecular Weight (g/mol): 248.39
Molecular Formula: C17H25F
InChi Key: VCIVJVKSOMDCMN-UHFFFAOYSA-N
SMILES: CCCCCC1CCC(CC1)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results