accessibility menu, dialog, popup

UserName

CAS RN 768-60-5

IUPAC Name: 1-ethynyl-4-methoxybenzene
Synonyms: 1-ethynyl-4-methoxybenzene
Molecular Weight (g/mol): 132.16
Molecular Formula: C9H8O
InChi Key: KBIAVTUACPKPFJ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C#C
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)