missing translation for 'onlineSavingsMsg'
Learn More

4-Ethynylanisole 98.0+%, TCI America™

Catalog No. E06035G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06035G 5 g
E06031G 1 g
E060325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E06035G Supplier TCI America Supplier No. E06035G

Chemical Identifiers

CAS 768-60-5
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.162
MDL Number MFCD00168815
InChI Key KBIAVTUACPKPFJ-UHFFFAOYSA-N
Synonym 4-ethynylanisole, 4-methoxyphenylacetylene, 4'-methoxyphenyl acetylene, 1-ethynyl-4-methoxy-benzene, 1-eth-1-ynyl-4-methoxybenzene, benzene, 1-ethynyl-4-methoxy, 4-methoxyphenyl acetylene, 4-ethynyl-1-methoxybenzene, 4'-methoxyphenylacetylene, p-ethynylanisole
PubChem CID 251020
IUPAC Name 1-ethynyl-4-methoxybenzene
SMILES COC1=CC=C(C=C1)C#C

Specifications

Color White-Yellow
Boiling Point 86°C
Quantity 5 g
Formula Weight 132.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Ethynylanisole
TSCA No
Recommended Storage Refrigerator