missing translation for 'onlineSavingsMsg'
Learn More

4-Ethynylanisole, 97%, Thermo Scientific™

Numéro de catalogue. p-3757910
Modifier l'affichage
Click to view available options
Quantité:
1g
5g
Emballage:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC379920010 1g Glass bottle
AC379920050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC379920010 Fournisseur Thermo Scientific Chemicals Code fournisseur 379920010

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 768-60-5
Formule moléculaire C9H8O
Poids moléculaire (g/mol) 132.16
Numéro MDL MFCD00168815
Clé InChI KBIAVTUACPKPFJ-UHFFFAOYSA-N
Synonyme 4-ethynylanisole, 4-methoxyphenylacetylene, 4'-methoxyphenyl acetylene, 1-ethynyl-4-methoxy-benzene, 1-eth-1-ynyl-4-methoxybenzene, benzene, 1-ethynyl-4-methoxy, 4-methoxyphenyl acetylene, 4-ethynyl-1-methoxybenzene, 4'-methoxyphenylacetylene, p-ethynylanisole
PubChem CID 251020
Nom de l’IUPAC 1-ethynyl-4-methoxybenzene
SOURIRES COC1=CC=C(C=C1)C#C

Spécifications

Nom ou substance chimique 4-Ethynylanisole
CAS % minimum 96.0
CAS % maximal 100.0
Densité 1.0190g/mL
Point d’ébullition 87°C to 91°C (11.0mmHg)
Point d’éclair 82°C
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 96% min. (GC)
Emballage Glass bottle
Formule linéaire HC∽C6H4OCH3
Indice de réfraction 1.5625 to 1.5675 (20°C, 589nm)
Quantité 1g
Gravité spécifique 1.019
Renseignements sur la solubilité Solubility in water: insoluble
Poids de formule 132.16
Pourcentage de pureté 97%
Afficher plus de résultats Afficher moins de résultats
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only