accessibility menu, dialog, popup

UserName

CAS RN 766-90-5

H3C(Z)

CAS RN 766-90-5

IUPAC Name: [(1Z)-prop-1-en-1-yl]benzene
Synonyms: (1Z)-prop-1-en-1-ylbenzene
Molecular Weight (g/mol): 118.18
Molecular Formula: C9H10
InChi Key: QROGIFZRVHSFLM-KXFIGUGUSA-N
SMILES: C\C=C/C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results