accessibility menu, dialog, popup

UserName

CAS RN 765-03-7

IUPAC Name: dodec-1-yne
Synonyms: 1-dodecyne
Molecular Weight (g/mol): 166.31
Molecular Formula: C12H22
InChi Key: ZVDBUOGYYYNMQI-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC#C
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)