missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecyne 95.0+%, TCI America™

Catalog No. D09975ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D09975ML 5 mL
D099725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D09975ML Supplier TCI America Supplier No. D09975ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 765-03-7
Molecular Formula C12H22
Molecular Weight (g/mol) 166.31
MDL Number MFCD00008960
InChI Key ZVDBUOGYYYNMQI-UHFFFAOYSA-N
Synonym 1-dodecyne, dodecyne, dodec-1-in, 1dodecyne, dodec-1-yn, 1-dodecyne 5g
PubChem CID 69821
IUPAC Name dodec-1-yne
SMILES CCCCCCCCCCC#C

Specifications

Color Yellow
Boiling Point 100°C
Quantity 5 mL
Formula Weight 166.31
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Dodecyne