accessibility menu, dialog, popup

UserName

KOHSOOO

CAS RN 7646-93-7

IUPAC Name: potassium hydrogen sulfate
Synonyms: potassium bisulfate
Molecular Weight (g/mol): 136.16
Molecular Formula: HKO4S
InChi Key: CHKVPAROMQMJNQ-UHFFFAOYSA-M
SMILES: [K+].OS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (9)
  • (5)
Percent Purity 
  • (1)
  • (2)
  • (5)
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
Grade 
  • (2)
  • (7)
  • (1)
Physical Form 
  • (2)
  • (5)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (2)
  • (5)
  • (6)
  • (9)
  • (5)
  • (1)
  • (2)
  • (5)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (7)
  • (1)
  • (2)
  • (5)
  • (3)