missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Bisulfate, Crystal, 35.0-37.0%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Crystals or Needles |
| Quantity | 45 kg |
| Molecular Weight (g/mol) | 136.17 |
| Percent Purity | 35 to 37% |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Chemical Name or Material | Potassium Bisulfate, Crystal |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.