accessibility menu, dialog, popup

UserName

H2COOOCH2

CAS RN 764-99-8

IUPAC Name: {2-[2-(ethenyloxy)ethoxy]ethoxy}ethene
Synonyms: {2-[2-(ethenyloxy)ethoxy]ethoxy}ethene
Molecular Weight (g/mol): 158.2
Molecular Formula: C8H14O3
InChi Key: SAMJGBVVQUEMGC-UHFFFAOYSA-N
SMILES: C=COCCOCCOC=C
Boiling Point 
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)