missing translation for 'onlineSavingsMsg'
Learn More

Di(ethylene glycol) divinyl ether, 95%

Catalog No. AC395410500
Change view
Click to view available options
Quantity:
50 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC395410500 50 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC395410500 Supplier Thermo Scientific Chemicals Supplier No. 395410500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 764-99-8 | C8H14O3 | 158.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 764-99-8
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.19
InChI Key SAMJGBVVQUEMGC-UHFFFAOYSA-N
Synonym diethylene glycol divinyl ether, divinylcarbitol, dvedeg, dvedeg russian, ether, bis 2-vinyloxy ethyl, ethene, 1,1'-oxybis 2,1-ethanediyloxy bis, 3,6,9-trioxaundeca-1,10-diene, unii-abm90y2g95, divinyl ether diethylenglykolu, bis 2-vinyloxy ethyl ether
PubChem CID 12998
IUPAC Name 1-ethenoxy-2-(2-ethenoxyethoxy)ethane
SMILES C=COCCOCCOC=C

Specifications

Melting Point -21.0°C
Density 0.9700g/mL
Boiling Point 168.0°C
Flash Point 88°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH2CHOCH2CH2OCH2CH2OCHCH2
Quantity 50 mL
Specific Gravity 0.97
Solubility Information Solubility in water: insoluble.
Formula Weight 158.19
Percent Purity 95%
Chemical Name or Material Di(ethylene glycol) divinyl ether
Show More Show Less
EINECSNumber 212-133-3

RUO – Research Use Only