missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Divinyl Ether (stabilized with KOH) 98.0+%, TCI America™

Catalog No. D049825ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D049825ML 25 mL
D0498100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D049825ML Supplier TCI America Supplier No. D049825ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 764-99-8
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00054610
InChI Key SAMJGBVVQUEMGC-UHFFFAOYSA-N
Synonym diethylene glycol divinyl ether, divinylcarbitol, dvedeg, dvedeg russian, ether, bis 2-vinyloxy ethyl, ethene, 1,1'-oxybis 2,1-ethanediyloxy bis, 3,6,9-trioxaundeca-1,10-diene, unii-abm90y2g95, divinyl ether diethylenglykolu, bis 2-vinyloxy ethyl ether
PubChem CID 12998
IUPAC Name 1-ethenoxy-2-(2-ethenoxyethoxy)ethane
SMILES C=COCCOCCOC=C

Specifications

Color Colorless
Boiling Point 197°C
Quantity 25 mL
Formula Weight 158.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Divinyl Ether (stabilized with KOH)
EINECSNumber (2)-2433&(7)-1321
RTECSNumber KN3850000
TSCA Yes