CAS RN 76262-87-8
CAS RN 76262-87-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Titanocene Bis(trifluoromethanesulfonate) 97.0+%, TCI America™
CAS: 76262-87-8 Molecular Formula: C12H10F6O6S2Ti MDL Number: MFCD00192505 Synonym: Bis(cyclopentadienyl)titanium(IV) Bis(trifluoromethanesulfonate)