missing translation for 'onlineSavingsMsg'
Learn More

Titanocene Bis(trifluoromethanesulfonate) 97.0+%, TCI America™

Catalog No. T1658100MG
Change view
Click to view available options
Quantity:
100 mg
Catalog No. Quantity
T1658100MG 100 mg
1 options

Catalog No. T1658100MG

Supplier: TCI America T1658100MG

Only null left

Chemical Identifiers

CAS 76262-87-8
Molecular Formula C12H10F6O6S2Ti
MDL Number MFCD00192505
Synonym Bis(cyclopentadienyl)titanium(IV) Bis(trifluoromethanesulfonate)

Specifications

Quantity 100 mg
Formula Weight 476.18
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Titanocene Bis(trifluoromethanesulfonate)