accessibility menu, dialog, popup

UserName

CAS RN 7620-28-2

IUPAC Name: (2R)-2-aminohexanedioic acid
Synonyms: D-2-aminoadipic acid
Molecular Weight (g/mol): 161.16
Molecular Formula: C6H11NO4
InChi Key: OYIFNHCXNCRBQI-SCSAIBSYSA-N
SMILES: N[C@H](CCCC(O)=O)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)