missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-2-Aminoadipic Acid 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 187°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 161.16 |
| Percent Purity | ≥95.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | D-2-Aminoadipic Acid |
Safety and Handling
| TSCA | No |