missing translation for 'onlineSavingsMsg'
Learn More

D-2-Aminoadipic acid, 98%, Thermo Scientific™

Catalog No. AC441960050
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC441960050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC441960050 Supplier Thermo Scientific Chemicals Supplier No. 441960050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7620-28-2
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.16
InChI Key OYIFNHCXNCRBQI-SCSAIBSYSA-N
Synonym d-2-aminoadipic acid, r-2-aminohexanedioic acid, 2r-2-aminohexanedioic acid, d-a-aminoadipic acid, d-, a-aminoadipic acid, r-2-aminoadipic acid, r-2-amino-hexanedioic acid, hexanedioic acid, 2-amino-,--and +, hexanedioic acid, 2-amino-, 2r, d-homoglutamic acid
PubChem CID 165627
ChEBI CHEBI:37025
IUPAC Name (2R)-2-aminohexanedioic acid
SMILES C(CC(C(=O)O)N)CC(=O)O

Specifications

Elemental Analysis 43.60 to 45.84% C, 8.47 to 8.91% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 5g
Specific Rotation -20.5
Formula Weight 161.16
Percent Purity 98%
Chemical Name or Material D-2-Aminoadipic acid

RUO – Research Use Only