accessibility menu, dialog, popup

UserName

CAS RN 761-65-9

IUPAC Name: N,N-dibutylformamide
Synonyms: formamide, N,N-dibutyl-
Molecular Weight (g/mol): 157.26
Molecular Formula: C9H19NO
InChi Key: NZMAJUHVSZBJHL-UHFFFAOYSA-N
SMILES: CCCCN(CCCC)C=O
Boiling Point 
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)