missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dibutylformamide, 99+%, Thermo Scientific Chemicals

Catalog No. AC112985000
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112985000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC112985000 Supplier Thermo Scientific Chemicals Supplier No. 112985000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 761-65-9 | Molecular Formula: C9H19NO | Molecular Weight: 157.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 761-65-9
Molecular Formula C9H19NO
Molecular Weight (g/mol) 157.26
MDL Number MFCD00003289
InChI Key NZMAJUHVSZBJHL-UHFFFAOYSA-N
Synonym dibutylformamide, formamide, n,n-dibutyl, n,n-di-n-butylformamide, formamide, n,n-di-n-butyl, dibutylamid kyseliny mravenci, dibutylamid kyseliny mravenci czech, formamide, dibutyl, n-formyldibutylamine, formamide,n-dibutyl, n,n-di-butylformamide
PubChem CID 12975
IUPAC Name N,N-dibutylformamide
SMILES CCCCN(CCCC)C=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Density 0.8600g/mL
Boiling Point 120.0°C (15.0 mmHg)
Flash Point 100°C
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.4430 to 1.4450
Linear Formula HCON[(CH2)3CH3]2
Quantity 500 mL
Beilstein 03,443F
Specific Gravity 0.86
Formula Weight 157.26
Percent Purity 99+%
Chemical Name or Material N, N-Dibutylformamide, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses
EINECSNumber 212-090-

RUO – Research Use Only